Skip to Content
MilliporeSigma
All Photos(5)

Documents

292664

Sigma-Aldrich

N-tert-Butyl-α-(2-sulfophenyl)nitrone sodium salt

95%

Synonym(s):

2-SPBN sodium salt, 2-Sulfonatophenyl tert-butyl nitrone sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SC6H4CH=N(O)C(CH3)3
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

95%

form

powder or crystals

mp

246 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CC(C)(C)[N+](\[O-])=C\c1ccccc1S([O-])(=O)=O

InChI

1S/C11H15NO4S.Na/c1-11(2,3)12(13)8-9-6-4-5-7-10(9)17(14,15)16;/h4-8H,1-3H3,(H,14,15,16);/q;+1/p-1/b12-8-;

InChI key

LAIQBHGUFDAJKL-JCTPKUEWSA-M

Application

Water-soluble, negatively-charged spin trap used to monitor aqueous phase of SDS micelles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Molnar et al.
Upsala journal of medical sciences, 115(3), 163-168 (2010-07-20)
Hyperglycemia exacerbates focal ischemic brain damage supposedly through various mechanisms. One such mechanism is oxidative stress involving reactive oxygen and nitrogen species (RONS) production. Nitrones attenuate oxidative stress in various models of brain injury. Sodium 2-sulfophenyl-N-tert-butyl nitrone (S-PBN) can be
G J Wilde et al.
Journal of neurochemistry, 69(2), 883-886 (1997-08-01)
The relative roles of the superoxide and hydroxyl radicals in oxidative stress-induced neuronal damage were investigated using organotypic hippocampal slice cultures. Cultures exposed to 100 microM duroquinone, a superoxide-generating compound, for 3 h developed CA1-selective lesions over a period of
Catherine E Hobbs et al.
Pediatrics international : official journal of the Japan Pediatric Society, 50(4), 481-488 (2008-10-22)
The production of oxygen free radicals after perinatal hypoxia-ischemia is thought to play a critical role in the pathogenesis of the brain injury. Administration of anti-oxidants may thus be neuroprotective. The aim of the present study was to investigate the
Steven L Peterson et al.
Neurotoxicology, 26(6), 969-979 (2005-05-14)
The status epilepticus (SE) induced in rats by lithium-pilocarpine (Li-pilo) shares many common features with soman-induced SE including extensive limbic neuropathology. Reactive oxygen species are hypothesized to play a role in the SE induced neuropathology and we propose that the
O A Andreassen et al.
Neuroreport, 12(15), 3371-3373 (2001-11-17)
There is substantial evidence implicating excitotoxicity and oxidative damage in the pathogenesis of Huntington's disease (HD). We therefore examined whether the antioxidants 2-sulpho-tert-phenyibutyinitrone (S-PBN) and alpha-lipoic acid could exert neuroprotective effects in transgenic mouse models of HD. S-PBN showed no

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service