Skip to Content
MilliporeSigma
All Photos(1)

Documents

287075

Sigma-Aldrich

Propidium iodide

95%

Synonym(s):

3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
Beilstein:
3843838
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95%

mp

220-225 °C (dec.) (lit.)

λmax

493 nm

SMILES string

[I-].[I-].CC[N+](C)(CC)CCC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C27H33N4.2HI/c1-4-31(3,5-2)17-9-16-30-26-19-22(29)13-15-24(26)23-14-12-21(28)18-25(23)27(30)20-10-7-6-8-11-20;;/h6-8,10-15,18-19,29H,4-5,9,16-17,28H2,1-3H3;2*1H/q+1;;/p-1

InChI key

XJMOSONTPMZWPB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manujendra N Saha et al.
PloS one, 7(1), e30215-e30215 (2012-01-26)
The low frequency of p53 alterations e.g., mutations/deletions (∼10%) in multiple myeloma (MM) makes this tumor type an ideal candidate for p53-targeted therapies. RITA is a small molecule which can induce apoptosis in tumor cells by activating the p53 pathway.
Ian K Campbell et al.
Journal of immunology (Baltimore, Md. : 1950), 192(11), 5031-5038 (2014-04-25)
High-dose i.v. Ig (IVIG) is used to treat various autoimmune and inflammatory diseases; however, the mechanism of action remains unclear. Based on the K/BxN serum transfer arthritis model in mice, IVIG suppression of inflammation has been attributed to a mechanism
Seiichiro Kobayashi et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2851-2861 (2014-04-15)
Cell adhesion molecule 1 (CADM1), initially identified as a tumor suppressor gene, has recently been reported to be ectopically expressed in primary adult T-cell leukemia-lymphoma (ATL) cells. We incorporated CADM1 into flow-cytometric analysis to reveal oncogenic mechanisms in human T-cell
Marcella Giovanni et al.
Journal of hazardous materials, 297, 146-152 (2015-05-10)
To cellular systems, nanoparticles are considered as foreign particles. Upon particles and cells contact, innate immune system responds by activating the inflammatory pathway. However, excessive inflammation had been linked to various diseases ranging from allergic responses to cancer. Common nanoparticles
Ramya Kumareswaran et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 116(3), 388-391 (2015-04-07)
Solid tumors contain regions of chronic and cyclic hypoxia. Chronic hypoxia can downregulate RAD51 and sensitize cells to PARP inhibition. Herein, we show that RAD51 expression, cell survival and toxicity to PARP inhibition is not affected under cyclic hypoxic conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service