Skip to Content
MilliporeSigma
All Photos(1)

Documents

28635-U

Hamilton® 1710N syringe

1710N Slim Line, volume 100 μL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

blunt tip stainless steel needle (point style 3)
glass body

needle size

22 ga (blunt tip)

description

gastight

feature

point style blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 203235

needle L

51 mm (2 in.)

volume

100 μL

compatibility

for use with CTC HTS PAL Instruments (HPLC Sampling)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service