Skip to Content
MilliporeSigma
All Photos(1)

Documents

27725

Sigma-Aldrich

Oxalic acid dihydrate

puriss., ≥99%

Synonym(s):

Ethanedioic acid dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCO2H · 2H2O
CAS Number:
Molecular Weight:
126.07
Beilstein:
3679436
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

grade

puriss.

Assay

≥99%

impurities

≤0.001% heavy metals (as Pb)

ign. residue

≤0.05% (as SO4)

mp

104-106 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤10 mg/kg

SMILES string

[H]O[H].[H]O[H].OC(=O)C(O)=O

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Paul A Nakata
PloS one, 10(10), e0141982-e0141982 (2015-10-31)
The establishment of new approaches to control chewing insects has been sought not only for direct use in reducing crop loss but also in managing resistance to the pesticides already in use. Engineered formation of calcium oxalate crystals is a
Radomír Pilný et al.
Dalton transactions (Cambridge, England : 2003), 43(32), 12243-12250 (2014-06-11)
Complex formation between [Pd(H2O)4](2+) and oxalate (ox = C2O4(2-)) has been studied spectrophoto-metrically in aqueous solution at variable temperature, ionic strength and pH. Thermodynamic parameters at 298.2 K and 1.00 mol dm(-3) HClO4 ionic medium for the complex formation [Pd(H2O)4](2+)
Jun Hu et al.
Physical chemistry chemical physics : PCCP, 17(32), 20537-20542 (2015-07-23)
Olive-shaped SnO2 nanocrystals were synthesized successfully via a facile hydrothermal route, using tin dichloride hydrate, oxalic acid dihydrate and polyvinylpyrrolidone as reaction precursors, and showed great potential in the large-scale preparation of SnO2 nanocrystals. The prepared SnO2 nanocrystals were characterized
Carina S D Wink et al.
Analytical and bioanalytical chemistry, 407(6), 1545-1557 (2015-01-13)
Lefetamine (N,N-dimethyl-1,2-diphenylethylamine, L-SPA) was marketed as an opioid analgesic in Japan and Italy. After being widely abused, it became a controlled substance. It seems to be a pharmaceutical lead for designer drugs because N-ethyl-1,2-diphenylethylamine (NEDPA) and N-iso-propyl-1,2-diphenylethylamine (NPDPA) were confiscated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service