Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27504

Sigma-Aldrich

L-(+)-Tartaric acid

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E334, 99.7-100.5% (calc. to the dried substance), grit

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

5.18 (vs air)

grade

puriss.

Assay

99.7-100.5% (calc. to the dried substance)

form

grit

optical activity

[α]20/D +12.0 to +12.8°, c = 20% in H2O

autoignition temp.

797 °F

quality

meets analytical specification of Ph. Eur., BP, NF, FCC, E334

impurities

organic volatile impurities, complies (GC)
residual solvents, complies
≤0.001% heavy metals (as Pb)

ign. residue

≤0.05% (as SO4)

loss

≤0.2% loss on drying, 105 °C

mp

168-170 °C
170-172 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
oxalate (C2O42-): in accordance
oxalate (as oxalic acid): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤200 mg/kg
Cu: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

application(s)

pharmaceutical (small molecule)

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
Preparation of organotrifluoroborate salts: precipitation-driven equilibrium under non-etching conditions.
Alastair J J Lennox et al.
Angewandte Chemie (International ed. in English), 51(37), 9385-9388 (2012-08-21)
Yanfang Feng et al.
Bioresource technology, 125, 138-144 (2012-10-03)
The aim of this study was to develop a promising and competitive bioadsorbent with the abundant of source, low price and environmentally friendly characters to remove cationic dye from wastewater. The swede rape straw (Brassica napus L.) modified by tartaric
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service