Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27420

Sigma-Aldrich

Citraconic acid

purum, ≥99.0% (T)

Synonym(s):

Methylmaleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
130.10
Beilstein:
1722679
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥99.0% (T)

ign. residue

≤0.1%

mp

88-94 °C (lit.)
91-95 °C

SMILES string

C\C(=C\C(O)=O)C(O)=O

InChI

1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2-

InChI key

HNEGQIOMVPPMNR-IHWYPQMZSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Ludmerczki et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(51), 11963-11974 (2019-06-30)
Thermal decomposition of citric acid is one of the most common synthesis methods for fluorescent carbon dots; the reaction pathway is, however, quite complex and the details are still far from being understood. For instance, several intermediates form during the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service