Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27319

Sigma-Aldrich

Reactive Blue 2 trisodium salt

Synonym(s):

1-Amino-4-[[4-[[4-chloro-6-[[3 (or 4)-sulfophenyl]amino]-1,3,5-triazin-2-yl]amino]-3-sulfophenyl]amino]-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid trisodium salt, Basilen Blue E-3G, Cibacron Blue F3GA, GST Inhibitor-1, Procion® Blue H-B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H17ClN7Na3O11S3
CAS Number:
Molecular Weight:
840.10
Colour Index Number:
61211
Beilstein:
6561550
EC Number:
UNSPSC Code:
12171500

Other Notes

A sulfonated triazine dye which can be used for probing nucleotide binding sites in numerous proteins ; Can be immobilized on a support matrix and used for affinity chromatography of proteins

Legal Information

Procion is a registered trademark of Dystar Textilfarben GmbH & Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Shen et al.
Chromatography, 21, 221-221 (2000)
A.T. Andrews et al.
Biotechnology Letters, 22, 1349-1349 (2000)
R.J. Ansell, C.R. Lowe
Applied Microbiology and Biotechnology, 51, 703-703 (1999)
S Subramanian
CRC critical reviews in biochemistry, 16(2), 169-205 (1984-01-01)
The dye Cibacron Blue F3GA has a high affinity for many proteins and enzymes. It has therefore been attached to various solid supports such as Sephadex, Sepharose, polyacrylamide, and the like. In the immobilized form the dye has rapidly been
J. Travis et al.
Methods in Enzymology, 80, 754-754 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service