Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27130

Sigma-Aldrich

Chromotrope 2B

for microscopy

Synonym(s):

2-(4-Nitrophenylazo)chromotropic acid disodium salt, Acid Red 176

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$45.70

$45.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$45.70

About This Item

Empirical Formula (Hill Notation):
C16H9N3Na2O10S2
CAS Number:
Molecular Weight:
513.37
Colour Index Number:
16575
Beilstein:
4109733
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

$45.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

for microscopy

Quality Level

form

powder or crystals

mp

300 °C

solubility

water: 10 mg/mL

ε (extinction coefficient)

≥500 at 509-519 nm in water

suitability

suitable for microscopy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1cc(cc2cc(c(\N=N\c3ccc(cc3)[N+]([O-])=O)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H11N3O10S2.2Na/c20-12-7-11(30(24,25)26)5-8-6-13(31(27,28)29)15(16(21)14(8)12)18-17-9-1-3-10(4-2-9)19(22)23;;/h1-7,20-21H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b18-17+;;

InChI key

AXUUHWJXDWBCSG-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chromotrope 2B along with Chromotrope 2R, was used to give a highly colored radical anion, during the spectrophotometric determination of cephalosporins (cephadroxil (I), cephalexin (II) and cephradine (III)) in pharmaceutical products.[1] It may be immobilized on AG 1-X8 ion exchange resin, used in flame atomic absorption spectrometry and inductively coupled plasma mass spectrometry (ICP-MS), done for the on-line preconcentration and determination of aluminium.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric assay of cephalosporins in pharmaceutical products, using chromotrope 2B and chromotrope 2R.
Issa YM and Amin AS
Microchimica Acta, 124 (3-4), 203-209 (1996)
On-line preconcentration of aluminium with immobilized Chromotrope 2B for the determination by flame atomic absorption spectrometry and inductively coupled plasma mass spectrometry.
Martin-esteban A
Analytica Chimica Acta, 304 (1), 121-126 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service