Key Documents
26156
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800
for use with Hamilton 1725 (LT, N, RN, or TLL), pkg of 1 ea
Select a Size
1 EA
$45.60
$45.60
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$45.60
About This Item
UNSPSC Code:
41122001
$45.60
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE plunger tip
size
250 μL
feature
1725LT, N, RN, or TLL
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 1162-03
compatibility
for use with Hamilton 1725 (LT, N, RN, or TLL)
General description
For field repair of Hamilton Series 1000, 1700 and 1800 syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)