Skip to Content
MilliporeSigma
All Photos(1)

Documents

25903

Sigma-Aldrich

(R)-(−)-2-Chloro-1-phenylethanol

purum, ≥97.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(−)-α-(Chloromethyl)benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH2Cl)OH
CAS Number:
Molecular Weight:
156.61
Beilstein:
4306747
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

purum

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D −48±2°, c = 2.8% in cyclohexane

optical purity

enantiomeric ratio: ≥99:1 (GC)

refractive index

n20/D 1.550
n20/D 1.552 (lit.)

density

1.185 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@@H](CCl)c1ccccc1

InChI

1S/C8H9ClO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6H2/t8-/m0/s1

InChI key

XWCQSILTDPAWDP-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral building block, used for the preparation of R(+)-phenyl-oxirane

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.J. Corey et al.
The Journal of Organic Chemistry, 53, 2861-2861 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service