Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24908

Sigma-Aldrich

6-Chloro-3-indolyl phosphate p-toluidine salt

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClNO4P · C7H9N
CAS Number:
Molecular Weight:
354.73
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98.0% (HPLC)

storage temp.

2-8°C

SMILES string

Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3cc(Cl)ccc23

InChI

1S/C8H7ClNO4P.C7H9N/c9-5-1-2-6-7(3-5)10-4-8(6)14-15(11,12)13;1-6-2-4-7(8)5-3-6/h1-4,10H,(H2,11,12,13);2-5H,8H2,1H3

InChI key

IUBOLLVEZYFKQN-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service