Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24549

Sigma-Aldrich

Petroleum ether

puriss., low boiling point hydrogen treated naphtha, bp 30-50 °C

Synonym(s):

Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
bp:
30-50 °C
Pricing and availability is not currently available.

grade

puriss.

form

liquid

impurities

≤0.005% non-volatile matter
≤0.01% S-compounds (as S)
≤0.02% benzene (GC)
≤0.02% water (Karl Fischer)

refractive index

n20/D 1.357-1.360

bp

30-50 °C

density

0.627-0.630 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is a low boiling point fraction having a boiling point in the range of 30-50°C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory, analgesic and antipyretic activities.[1]

Application

Petroleum ether may be used in the absorption chromatographic purification of organic compounds.[2]

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Pertains only in Germany: unversteuert

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid chromatographic technique for preparative separations with moderate resolution.
Still WC, et al.
The Journal of Organic Chemistry, 43(14), 2923-2925 (1978)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service