Documents
24530
Hamilton® syringe, luer tip
701LT, volume 10 μL, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80301
needle L
, n/a
volume
10 μL
compatibility
for use with Hamilton 701LT (Luer tip)
General description
Hamilton® syringe, luer tip is, ideal for dispensing volume from 0.5μL up to 500μL. It features a ground glass luer tip suitable for mounting chromatographic and PTFE needles.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional, 54-54 (2009)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192-192 (2012)