Skip to Content
MilliporeSigma
All Photos(1)

Documents

24390

Sigma-Aldrich

Chloromethyl methyl sulfide

technical, ≥80% (GC)

Synonym(s):

Chlorodimethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2Cl
CAS Number:
Molecular Weight:
96.58
Beilstein:
1730851
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

technical

Assay

≥80% (GC)

refractive index

n20/D 1.498 (lit.)
n20/D 1.498

bp

105 °C (lit.)

density

1.153 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CSCCl

InChI

1S/C2H5ClS/c1-4-2-3/h2H2,1H3

InChI key

JWMLCCRPDOIBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the introduction of the methylthiomethyl (MTM) protecting group into carboxylic acids. MTM esters are stable to mild reducing agents

replaced by

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.G. Wade et al.
Tetrahedron Letters, 731-731 (1978)
[Mutagenic activity of chlorodimethylsulfide and some of its aniline derivatives].
M Vito et al.
Bollettino della Societa italiana di biologia sperimentale, 61(6), 917-923 (1985-06-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service