Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24320

Sigma-Aldrich

2-Chloro-4,6-dimethoxy-1,3,5-triazine

purum, ≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6ClN3O2
CAS Number:
Molecular Weight:
175.57
Beilstein:
148988
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (AT)

mp

71-74 °C (lit.)
75-79 °C

SMILES string

COc1nc(Cl)nc(OC)n1

InChI

1S/C5H6ClN3O2/c1-10-4-7-3(6)8-5(9-4)11-2/h1-2H3

InChI key

GPIQOFWTZXXOOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Intermediate for the synthesis of triazines. Activating agent for peptide coupling; Direct reduction of active esters to aldehydes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple Method for the Reduction of Carboxylic Acids to Aldehydes or Alcohols Using H(2) and Pd/C.
Massimo Falorni et al.
The Journal of organic chemistry, 64(24), 8962-8964 (2001-10-25)
Z.J. Kaminski et al.
The Journal of Organic Chemistry, 63, 4248-4248 (1998)
Tatiana V Esipova et al.
Cell metabolism, 29(3), 736-744 (2019-01-29)
Quantitative imaging of oxygen distributions in tissue can provide invaluable information about metabolism in normal and diseased states. Two-photon phosphorescence lifetime microscopy (2PLM) has been developed to perform measurements of oxygen in vivo with micron-scale resolution in 3D; however, the method's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service