Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24020

Sigma-Aldrich

Ethyl 4-chlorobutyrate

purum, ≥97.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3CO2C2H5
CAS Number:
Molecular Weight:
150.60
Beilstein:
1098810
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.5% (GC)

refractive index

n20/D 1.432 (lit.)
n20/D 1.432

bp

186 °C (lit.)

density

1.075 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCl

InChI

1S/C6H11ClO2/c1-2-9-6(8)4-3-5-7/h2-5H2,1H3

InChI key

OPXNFHAILOHHFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace Thome et al.
American journal of physiology. Cell physiology, 317(4), C701-C713 (2019-07-11)
Chronic kidney disease (CKD) leads to increased skeletal muscle fatigue, weakness, and atrophy. Previous work has implicated mitochondria within the skeletal muscle as a mediator of muscle dysfunction in CKD; however, the mechanisms underlying mitochondrial dysfunction in CKD are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service