Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239852

Sigma-Aldrich

Benzophenone

ReagentPlus®, ≥99%

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein:
1238185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 108 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder

bp

305 °C (lit.)

mp

47-51 °C (lit.)

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire Chatalova-Sazepin et al.
Angewandte Chemie (International ed. in English), 54(18), 5443-5446 (2015-03-05)
A three-component carboetherification of unactivated alkenes has been developed allowing the rapid building of complexity from simple starting materials. A wide range of α-substituted styrenes underwent smooth reactions with unactivated alkyl nitriles and alcohols to afford γ-alkoxy alkyl nitriles with
Masayuki Kyomoto et al.
Biomaterials, 34(32), 7829-7839 (2013-07-31)
We investigated the production of free radicals on a poly(ether-ether-ketone) (PEEK) substrate under ultraviolet (UV) irradiation. The amount of the ketyl radicals produced from the benzophenone (BP) units in the PEEK molecular structure initially increased rapidly and then became almost
L L Barnkob et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 30(2), 395-402 (2012-11-17)
The aim of this study was to investigate the effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax®. Kinetic migration investigations were carried out with three relative humidities in the interval between 39%
Haidong Li et al.
Talanta, 99, 811-815 (2012-09-13)
Benzophenone is one of the most commonly used photoinitiators of UV-cured inks on food packaging materials and can migrate into foodstuffs. In this study, an amperometric benzophenone sensor based on molecularly imprinted polymer (MIP) was successfully constructed for the first
Feifan Yu et al.
PloS one, 8(2), e56597-e56597 (2013-02-21)
Affinity proteins binding to antibody constant regions have proved to be invaluable tools in biotechnology. Here, protein engineering was used to expand the repertoire of available immunoglobulin binding proteins via improvement of the binding strength between the widely used staphylococcal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service