Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

23568

Sigma-Aldrich

Chlorobenzene

puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

grade

absolute
puriss.

Assay

≥99.5% (GC)

autoignition temp.

1178 °F

quality

over molecular sieve (H2O ≤0.005%)

expl. lim.

7.1 %

impurities

≤0.005% water

refractive index

n20/D 1.524 (lit.)
n20/D 1.525

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coq B, et al.
J. Catal., 101(2), 434-445 (1986)
Sedlak DL and Andren AW.
Environmental Science & Technology, 25(4), 777-782 (1991)
T F Guerin et al.
Letters in applied microbiology, 32(5), 340-345 (2001-05-01)
The aim of the current study was to test whether resazurin changed colour when incubated with a range of organic chemicals used as growth substrates in bioremediation studies and to determine whether resazurin was more effective in estimating microbial growth
Yang J, et al.
Macromolecules, 41(24), 9650-9655 (2008)
Yiling Wang et al.
ACS applied materials & interfaces, 7(13), 7170-7179 (2015-03-21)
Solution-processed organic-inorganic hybrids composing of MoO3 nanoparticles and PEDOT:PSS were developed for use in inverted organic solar cells as hole transporting layer (HTL). The hybrid MoO3:PEDOT:PSS inks were prepared by simply mixing PEDOT:PSS aqueous and MoO3 ethanol suspension together. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service