Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23240

Sigma-Aldrich

4-Nitrophenyl chloroformate

purum, ≥97.0% (AT)

Synonym(s):

Chloroformic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C6H4NO2
CAS Number:
Molecular Weight:
201.56
Beilstein:
518127
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (AT)

bp

159-162 °C/19 mmHg (lit.)

mp

76-82 °C
77-79 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(OC(Cl)=O)cc1

InChI

1S/C7H4ClNO4/c8-7(10)13-6-3-1-5(2-4-6)9(11)12/h1-4H

InChI key

NXLNNXIXOYSCMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the synthesis of mixed, activated carbonates and thiocarbonates, activated urethanes, and other compounds[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Phillips et al.
Tetrahedron Letters, 31, 3291-3291 (1990)
Kai Dong et al.
International journal of nanomedicine, 11, 5109-5123 (2016-10-28)
In this study, a type of multifunctional mixed micelles were prepared by a novel biodegradable amphiphilic polymer (MPEG-SS-2SA) and a multidrug resistance (MDR) reversal agent (d-α-tocopheryl polyethylene glycol succinate, TPGS). The mixed micelles could achieve rapid intracellular drug release and
Zhengjie Yang et al.
Journal of orthopaedic translation, 21, 57-65 (2020-02-27)
The survival rate of osteosarcoma therapy still lags behind overall cancer therapies due to the intrinsic or acquired drug resistance. Developing novel drug delivery systems that may overcome drug resistance would greatly facilitate osteosarcoma therapy. Poly(ethylene glycol) (PEG)-sheddable reduction-sensitive polyurethane
Jingjing Pan et al.
ACS applied materials & interfaces, 6(16), 14391-14398 (2014-07-18)
In this work, a novel gene delivery strategy was proposed based on silicon nanowire arrays modified with high-molecular-weight 25 kDa branched polyethylenimine (SN-PEI). Both the plasmid DNA (pDNA) binding capacity and the in vitro gene transfection efficiency of silicon nanowire
Feng Ren et al.
Chemical & pharmaceutical bulletin, 62(9), 898-905 (2014-09-02)
A series of novel benzyloxyurea derivatives was designed, synthesized by substituting different benzyls or phenyls on N,N'-positions of the hydroxyurea (HU). These target compounds were evaluated for their anticancer activity in vitro against human leukemia cell line K562 and murine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service