Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23019

Sigma-Aldrich

2-Chloroethanesulfonyl chloride

purum, ≥97.0% (AT)

Synonym(s):

2-Chloro-1-ethanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2SO2Cl
CAS Number:
Molecular Weight:
163.02
Beilstein:
1751202
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor pressure

15 mmHg ( 84 °C)

grade

purum

Assay

≥97.0% (AT)

refractive index

n20/D 1.493 (lit.)
n20/D 1.494

bp

84-86 °C/15 mmHg (lit.)

density

1.56 g/mL at 25 °C (lit.)

SMILES string

ClCCS(Cl)(=O)=O

InChI

1S/C2H4Cl2O2S/c3-1-2-7(4,5)6/h1-2H2

InChI key

VHCSBTPOPKFYIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service