Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22126

Sigma-Aldrich

Cefsulodin sodium salt

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N4NaO8S2
CAS Number:
Molecular Weight:
554.53
EC Number:
MDL number:
UNSPSC Code:
51101500

Assay

≥98.0% (TLC)

impurities

≤3% water

solubility

H2O: 50 mg/mL, clear, light yellow

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12SCC(C[n+]3ccc(cc3)C(N)=O)=C(N1C(=O)[C@H]2NC(=O)C(c4ccccc4)S([O-])(=O)=O)C([O-])=O

General description

Chemical structure: ß-lactam

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service