Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21800

Sigma-Aldrich

Carbenicillin disodium salt

≥90% (anhydrous basis)

Synonym(s):

α-Carboxybenzylpenicillin disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2Na2O6S
CAS Number:
Molecular Weight:
422.36
Beilstein:
5722128
EC Number:
MDL number:
UNSPSC Code:
51101500
Pricing and availability is not currently available.

Assay

≥90% (anhydrous basis)

solubility

H2O: 50 mg/mL

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C17H18N2O6S.2Na/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8;;/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25);;/q;2*+1/p-2/t9?,10-,11+,14-;;/m1../s1

InChI key

RTYJTGSCYUUYAL-YCAHSCEMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Biochem/physiol Actions

Mode of Action: Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.


Antimicrobial spectrum: Active against Gram-positive and Gram-negative bacteria

Analysis Note

Stable at 37 °C for 3 days

Other Notes

Parenteral antibiotic active against Pseudomonas aeruginosa.; Characterization of two chromosome-encoded β-lactamases from Citrobacter diversus ULA-27[1]

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Amicosante et al.
The Biochemical journal, 254(3), 885-890 (1988-09-15)
Two chromosome-encoded beta-lactamases have been purified from Citrobacter diversus ULA-27. They exhibited slightly different isoelectric points (6.8 and 6.2) and very similar Mr values (congruent to 29,000). Their specificity spectrum was rather wide, since they hydrolysed some cephalosporins with kcat:
Edward Avilés et al.
Organic letters, 12(22), 5290-5293 (2010-10-23)
Monamphilectine A (1), a new diterpenoid β-lactam alkaloid showing potent antimalarial activity, was isolated in milligram quantities following bioassay-directed extraction of a Puerto Rican marine sponge Hymeniacidon sp. Its structure, established by interpretation of spectral data, was confirmed unequivocally by
Liang Yang et al.
Journal of microbiology (Seoul, Korea), 49(1), 107-114 (2011-03-04)
Resistance-Nodulation-Cell Division (RND) pumps play important roles in bacterial resistance to antibiotics. Pseudomonas aeruginosa is an important human pathogen which exhibits high level resistance to antibiotics. There are total of 12 RND pumps present in the P. aeruginosa PAOl genome.
Olivier Fisette et al.
Biophysical journal, 103(8), 1790-1801 (2012-10-23)
The effects of substrate binding on class A β-lactamase dynamics were studied using molecular dynamics simulations of two model enzymes; 40 100-ns trajectories of the free and substrate-bound forms of TEM-1 (with benzylpenicillin) and PSE-4 (with carbenicillin) were recorded (totaling
J Steinmann et al.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin, 16(33) (2011-08-30)
We describe the epidemiology and characteristics of the pathogen and patients (n=7) associated with an outbreak of a carbapenem-resistant Klebsiella pneumoniae (CRKP) strain in a German university hospital from July 2010 to January 2011. Species identification and detection of carbapenem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service