Skip to Content
MilliporeSigma
All Photos(1)

Documents

21795

Sigma-Aldrich

Carbazole-9-carbonyl chloride

purum, for fluorescence, ≥98.0% (GC)

Synonym(s):

9-(Chloroformyl)carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClNO
CAS Number:
Molecular Weight:
229.66
Beilstein:
161868
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence
purum

Assay

≥98.0% (GC)

mp

100-104 °C

storage temp.

2-8°C

SMILES string

ClC(=O)n1c2ccccc2c3ccccc13

Application

Reagent used for the fluorogenic labeling of amino acids.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Allenmark
Tetrahedron Letters, 31, 1455-1455 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service