Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

215651

Sigma-Aldrich

Disperse Blue 3

Dye content 20 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O3
CAS Number:
Molecular Weight:
296.32
Colour Index Number:
61505
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 20%

λmax

594 nm
640 nm (2nd)

SMILES string

CNc1ccc(NCCO)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C17H16N2O3/c1-18-12-6-7-13(19-8-9-20)15-14(12)16(21)10-4-2-3-5-11(10)17(15)22/h2-7,18-20H,8-9H2,1H3

InChI key

NLXFWUZKOOWWFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Blue 3 belongs to the class of anthraquinone dyes.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Kanerva, P. Elsner, J.E. Wahlberg, H.I. Maibach
Handbook of Occupational Dermatology (2013)
L. Kanerva, P. Elsner, J.E. Wahlberg, H.I. Maibach
Handbook of Occupational Dermatology (2013)
Davide Tamburro et al.
Journal of the American Chemical Society, 133(47), 19178-19188 (2011-10-18)
Many low-abundance biomarkers for early detection of cancer and other diseases are invisible to mass spectrometry because they exist in body fluids in very low concentrations, are masked by high-abundance proteins such as albumin and immunoglobulins, and are very labile.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service