Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21286

Supelco

(1R)-(+)-Camphanic chloride

for chiral derivatization, ≥97.0%

Synonym(s):

(1R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClO3
CAS Number:
Molecular Weight:
216.66
Beilstein:
4842302
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

grade

derivatization grade ((chiral))
for chiral derivatization

product line

ChiraSelect

Assay

≥97.0% (AT)
≥97.0%

form

solid

optical activity

[α]20/D +18±1°, c = 2% in carbon tetrachloride

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

mp

67-70 °C

storage temp.

−20°C

SMILES string

CC1(C)[C@]2(C)CC[C@]1(OC2=O)C(Cl)=O

InChI

1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m1/s1

InChI key

PAXWODJTHKJQDZ-ZJUUUORDSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service