Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

210331

Sigma-Aldrich

Acid Green 27

Dye content 65 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32N2Na2O8S2
CAS Number:
Molecular Weight:
706.74
Colour Index Number:
61580
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 65%

mp

258-260 °C (lit.)

λmax

605 nm
642 nm (2nd)

SMILES string

[Na+].[Na+].CCCCc1ccc(Nc2ccc(Nc3ccc(CCCC)cc3S([O-])(=O)=O)c4C(=O)c5ccccc5C(=O)c24)c(c1)S([O-])(=O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service