Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21023

Sigma-Aldrich

Cesium trifluoroacetate

BioUltra, ≥99.0% (NT)

Synonym(s):

CsTFA, Trifluoroacetic acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOCs
CAS Number:
Molecular Weight:
245.92
Beilstein:
4161420
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

product line

BioUltra

Assay

≥99.0% (NT)

form

solid

impurities

Insoluble matter, passes filter test

pH

9.0-10.5 (25 °C, 2 M in H2O)

mp

114-116 °C (lit.)

solubility

H2O: 2 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤10 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

2 M in H2O

UV absorption

λ: 260 nm Amax: 0.09
λ: 280 nm Amax: 0.04

SMILES string

[Cs+].[O-]C(=O)C(F)(F)F

InChI

1S/C2HF3O2.Cs/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

RJYSYRSELCQCSO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Isolation of proteoglycans using phenol extraction and isopycnic centrifugation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A McBain et al.
Analytical biochemistry, 188(1), 203-213 (1990-07-01)
A procedure for the rapid isolation of heparan sulfate proteoglycans from Zwittergent lysates of cultured cells and cetylpyridinium precipitates of culture medium is described. After removal of most nonglycosylated proteins by phenol extraction, the proteoglycans are fractionated and isolated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service