Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20944

Hamilton® syringe, 7000 series, knurled hub with guide

7001, volume 1 μL, needle size 25s ga (blunt tip), needle L 70 mm (2.75 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$180.00

$180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$180.00

About This Item

UNSPSC Code:
41122001

$180.00


Please contact Customer Service for Availability

Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)

needle size

25s ga (blunt tip)

feature

KH (Knurled Hub)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80107

needle L

70 mm (2.75 in.)

volume

1 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service