Skip to Content
MilliporeSigma
All Photos(1)

Documents

20838-U

Hamilton® syringe barrel

for use with Hamilton 801RN, volume 10 μL, pkg of 1 ea

Synonym(s):

Barrel Assembly for Hamilton® Syringe 801RN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

glass

size

10 μL

description

801RN

feature

graduations 0.2 μL

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 32129

volume

10 μL

compatibility

for use with Hamilton 801RN

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service