Skip to Content
MilliporeSigma
All Photos(1)

Documents

20800

Sigma-Aldrich

8-Butyryloxypyrene-1,3,6-trisulfonic acid trisodium salt

for fluorescence, ≥95.0% (TLC)

Synonym(s):

Trisodium 8-butyryloxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13Na3O11S3
CAS Number:
Molecular Weight:
594.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥95.0% (TLC)

mp

240-250 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble
methanol: soluble

fluorescence

λex 460 nm; λem 510 nm in 0.1 M Tris pH 8.0

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].CCCC(=O)Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Other Notes

Highly water soluble enzyme substrate for the fluorimetric assay of esterases at longwave excitation and emission wavelengths.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O S Wolfbeis et al.
Analytical biochemistry, 129(2), 365-370 (1983-03-01)
A direct and continuous kinetic method for the fluorimetric assay of various hydrolases by using new, highly water-soluble substrates is described. The latter consist of esters of strongly fluorescent 1-hydroxypyren-3,6,8-trisulfonic acid trisodium salt with acetic, butyric, caprylic, and oleic acid.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service