Key Documents
20721
Hamilton® syringe, 1000 series GASTIGHT®, luer tip
1002LT, volume 2.5 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81401
needle L
, n/a
volume
2.5 mL
Related Categories
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. is it possible to have the dimensions of the syringe?
    1 answer
    1. This is a product of Hamilton. The barrel inner diameter is 0.287 inches (7.29 mm) and the barrel outer diameter is 0.407 inches (10.3 mm). Please see the image below to review the complete product specifications.
      Helpful?