Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20270

Sigma-Aldrich

2-Hexanone

purum, ≥96.0% (GC)

Synonym(s):

Butyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
1737676
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
127 °C (lit.)
vapor pressure:
10 mmHg ( 39 °C)

vapor pressure

10 mmHg ( 39 °C)

grade

purum

Assay

≥96.0% (GC)

form

liquid

technique(s)

GC/GC: suitable

refractive index

n20/D 1.401 (lit.)
n20/D 1.401

bp

127 °C (lit.)

mp

−57 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

SMILES string

CCCCC(C)=O

InChI

1S/C6H12O/c1-3-4-5-6(2)7/h3-5H2,1-2H3

InChI key

QQZOPKMRPOGIEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile organic compounds in exhaled breath by ion mobility spectrometry.
Ulanowska A, et al.
Chem. Anal. (Warsaw), 53(6), 953-965 (2008)
Thermodynamic properties of binary mixtures of 2-hexanone withn-alkanes at 35? C.
Legido JL, et al.
Journal of Solution Chemistry, 19(11), 1095-1102 (1990)
Deuterium isotope effects in the photochemistry of 2-hexanone.
Coulson DR and Yang NC.
Journal of the American Chemical Society, 88(19), 4511-4513 (1966)
P Manini et al.
Rapid communications in mass spectrometry : RCM, 12(21), 1615-1624 (1998-11-10)
A liquid chromatography atmospheric pressure electrospray mass spectrometry (ESI-LC/MS) system was evaluated for the identification and characterization of n-hexane conjugated metabolites (glucuronides) in untreated urine samples. Chromatography of glucuronides was obtained under ion-suppressed reversed-phase conditions, by using high-speed (3 cm
Walter F Schmidt et al.
Chemistry and physics of lipids, 142(1-2), 23-32 (2006-04-18)
Stearic acid (SA) is highly soluble in structurally diverse solvents. SA/solvent packing within a (24.8 A)3 cubic volume explains the stoichiometry of SA solubility at multiple temperatures in multiple solvents. In the absence of solvent, the cubic volume contains 25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service