Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201227

Sigma-Aldrich

Fast Blue RR Salt

85%

Synonym(s):

4-Benzoylamino-2,5-dimethoxybenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 24

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H14ClN3O3 · 1/2 ZnCl2
CAS Number:
Molecular Weight:
387.89
Colour Index Number:
37155
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

85%

λmax

329 nm

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc([N+]#N)c(OC)cc1NC(=O)c2ccccc2.COc3cc([N+]#N)c(OC)cc3NC(=O)c4ccccc4

InChI

1S/2C15H13N3O3.4ClH.Zn/c2*1-20-13-9-12(18-16)14(21-2)8-11(13)17-15(19)10-6-4-3-5-7-10;;;;;/h2*3-9H,1-2H3;4*1H;/q;;;;;;+2/p-2

InChI key

LLARVKAOQLUCQI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service