Skip to Content
MilliporeSigma
All Photos(1)

Documents

19660

Supelco

4-tert-Butylbenzoyl chloride

purum, ≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4COCl
CAS Number:
Molecular Weight:
196.67
Beilstein:
775793
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

refractive index

n20/D 1.536 (lit.)
n20/D 1.536

bp

135 °C/20 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)C(Cl)=O

InChI

1S/C11H13ClO/c1-11(2,3)9-6-4-8(5-7-9)10(12)13/h4-7H,1-3H3

InChI key

WNLMYNASWOULQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service