Skip to Content
MilliporeSigma
All Photos(1)

Documents

19023

Supelco

N-Acetylneuraminic acid

analytical standard

Synonym(s):

N-Acetyl-D-neuraminic acid, N-acetyl-beta-neuraminic acid, N-acetylneuramic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid, Lactaminic acid, NAN, NANA, Sialic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO9
CAS Number:
Molecular Weight:
309.27
Beilstein:
1716283
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

O[C@@]1(O[C@@]([C@@H]([C@H](C1)O)NC(C)=O)([H])[C@@H]([C@@H](CO)O)O)C(O)=O

InChI

1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,7+,8+,9+,11-/m0/s1

InChI key

SQVRNKJHWKZAKO-PFQGKNLYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Digestibility and Microbiota Modulation by Dietary Sialic Acid: Research indicated that sialylation of dietary mucin can modulate its digestibility and subsequently affect the gut microbiota of elderly individuals, highlighting the significance of N-Acetylneuraminic acid in dietary formulations and gut health (Fang et al., 2024).



Biochem/physiol Actions

Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins. Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion. Membrane glycoproteins are also important in tumor growth and metastases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecularly imprinted polymers coated on the surface of metal--organic frameworks combined with liquid chromatography-tandem mass spectrometry for the detection of free N-acetylneuraminic acid in serum of pneumoconiosis patients
Lin R, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 181, 107821-107821 (2022)

Articles

Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service