Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18894

Supelco

2-Hydroxy-3-methoxybenzaldehyde thiosemicarbazone

for spectrophotometric determination of Ni2+, ≥99.0% (HPLC)

Synonym(s):

N-(2-Hydroxy-3-methoxybenzylidene)thiosemicarbazide, o-Vanillin thiosemicarbazone, HMBATSC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N3O2S
CAS Number:
Molecular Weight:
225.27
Beilstein:
2117346
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Assay

≥99.0% (HPLC)

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

COc1cccc(\C=N\NC(N)=S)c1O

InChI

1S/C9H11N3O2S/c1-14-7-4-2-3-6(8(7)13)5-11-12-9(10)15/h2-5,13H,1H3,(H3,10,12,15)/b11-5+

InChI key

ZOZTWVGHQKXQOZ-VZUCSPMQSA-N

General description

2-Hydroxy-3-methoxybenzaldehyde thiosemicarbazone (HMBATSC) can be prepared by dissolving 2-Hydroxy-3-methoxybenzaldehyde and thiosemicarbazide in methanol, followed by further filtration and recrystallization.

Application

HMBATSC may be used in the determination of molybdenum in aqueous dimethyl formamide medium using second derivative spectrophotometric method. It may be used in determination of nickel(II) using spectrophotometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct and second derivative spectrophotometric determination of molybdenum (VI) in food stuffs and in alloy steels using 2-hydroxy-3-methoxy benzaldehyde thiosemicarbazone (HMBATSC).
Praveenkumar, A., P. Raveendra Reddy, and V. Krishna Reddy.
Journal of the Chilean Chemical Society, 52, 1309-1313 (2007)
Spectrophotometric determination of nickel (II) with 2-hydroxy-3-methoxy benzaldehyde thiosemicarbazone.
Kumar, A. Praveen, P. Raveendra Reddy, and V. Krishna Reddy.
Indian J. Chem. A, 46, 1625-1629 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service