Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

178268

Sigma-Aldrich

4-(2-Pyridylazo)resorcinol monosodium salt hydrate

Synonym(s):

PAR monosodium salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$209.00
10 G
$461.00

$209.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$209.00
10 G
$461.00

About This Item

Empirical Formula (Hill Notation):
C11H8N3NaO2 · xH2O
CAS Number:
Molecular Weight:
237.19 (anhydrous basis)
Beilstein:
3825637
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$209.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.5% (HPLC)

Quality Level

form

powder or crystals

λmax

411 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[H]O[H].Oc1cc([O-])ccc1\N=N\c2ccccn2

InChI

1S/C11H9N3O2.Na.H2O/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;;/h1-7,15-16H;;1H2/q;+1;/p-1/b14-13+;;

InChI key

KMHFHNVYQMFVAN-QDBORUFSSA-M

General description

4-(2-Pyridylazo)resorcinol monosodium salt is a chromophoric indicator and chromogenic chelator employed for the spectrophotometric determination of several metal ions.

Application

4-(2-Pyridylazo)resorcinol monosodium salt hydrate has been used in the preparation of titanium reagent for the spectrometric measurement of hydrogen peroxide (H2O2) and superoxide anion (O2•-). It is also suitable to measure the concentration of Zn2+, Zinc transition metal ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Chemistry, 57, 625-625 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service