Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17798

Sigma-Aldrich

Pongamol

purum, ≥97.0% (TLC)

Synonym(s):

1-(4-Methoxy-5-benzofuranyl)-3-phenyl-1,3-propanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14O4
CAS Number:
Molecular Weight:
294.30
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥97.0% (TLC)

mp

126-130 °C

SMILES string

COc1c(ccc2occc12)C(=O)CC(=O)c3ccccc3

Looking for similar products? Visit Product Comparison Guide

Other Notes

Standard for determination of pongamol by reversed-phase HPLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V.K. Gore et al.
Analytical Letters, 33, 337-337 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service