Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16499

Supelco

Ipriflavone

analytical standard

Synonym(s):

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one, Ipriflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O3
CAS Number:
Molecular Weight:
280.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

116-120 °C (lit.)

format

neat

SMILES string

CC(C)Oc1ccc2c(OC=C(C2=O)c3ccccc3)c1

InChI

1S/C18H16O3/c1-12(2)21-14-8-9-15-17(10-14)20-11-16(18(15)19)13-6-4-3-5-7-13/h3-12H,1-2H3

InChI key

SFBODOKJTYAUCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ipriflavone, a flavonoid derivative of the natural isoflavone compound, plays a role in the mitochondrial phosphorylation and exhibits growth-promoting activity.

Application

Ipriflavone can be used as an analytical standard for the determination of the analyte in human and animal biological samples by liquid chromatography (LC) based methods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of ipriflavone and its main metabolites M1 and M5 in human plasma by liquid chromatography with tandem mass spectrometry
Ji HY, et al.
Analytical Letters, 38(1), 99-110 (2005)
Disposition of ipriflavone (TC-80) in rats and dogs.
Yoshida K, et al.
Radioisotopes, 34(11), 618-623 (1985)
Determination of ipriflavone and its synthetic impurities by high-performance liquid chromatography using diode-array detection
Sustacha K, et al.
Journal of Chromatography A, 719(1), 245-250 (1996)
H J Chung et al.
Research in veterinary science, 85(1), 149-155 (2007-10-09)
It has been reported that ipriflavone was primarily metabolized via hepatic microsomal cytochrome P450 (CYP) 1A1/2 and 2C11 in rats, and the expression of hepatic CYP1A2 and 2C11 was not changed in rats with water deprivation for 72h compared to
Pharmacokinetics of ipriflavone, an isoflavone derivative, after intravenous and oral administration to rats: hepatic and intestinal first-pass effects
Kim SH and Lee MG
Life Sciences, 70(11), 1299-1315 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service