Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16313

Supelco

Butyl isothiocyanate

analytical standard

Synonym(s):

Benzyl ITC, Butyl ITC, NSC 194808

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NCS
CAS Number:
Molecular Weight:
115.20
Beilstein:
906839
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.5 (lit.)

bp

70-71 °C/35 mmHg (lit.)

density

0.955 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCN=C=S

InChI

1S/C5H9NS/c1-2-3-4-6-5-7/h2-4H2,1H3

InChI key

LIMQQADUEULBSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl isothiocyanate, an organic isothiocyanate, is a common volatile constituent of various vegetables and plants.[1][2][3][4][5][6]

Application

Butyl isothiocyanate can be used as an analytical standard in the determination of the analyte by using the following techniques:
  • High-performance liquid chromatography (HPLC) determination of rapeseeds and rapeseed meal,[1] and enzyme hydrolysates of Tropaeolum tuberosum.[2]
  • Gas chromatography-mass spectrometry (GC-MS) analysis of Chinese medicinal herbs[3] and horseradish samples.[7]

It may also be used as an internal standard for the quantification of isothiocyanate irritant constituents in Cappers Spinoza by GC.[8]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of isothiocyanates, indoles, and oxazolidinethiones in myrosinase digests of rapeseeds and rapeseed meal by HPLC
Matthaus B and Fiebig HJ
Journal of Agricultural and Food Chemistry, 44(12), 3894-3899 (1996)
Isothiocyanates and thioureas in enzyme hydrolysates of Tropaeolum tuberosum
Johns T and Towers GHN
Phytochemistry, 20(12), 2687-2689 (1981)
Determination of Three Irritant Constituents of Cappers spinoza with Capillary Gas Chromatography by Using Internal Standard Method [J]
GUO XN, et al.
Chinese Journal of Experimental Medical Formulae, 16 (2012)
Fenaroli's Handbook of Flavor Ingredients, 72(5), 307-309 (2019)
SPME-GC-MS ANALYSIS OF HORSERADISH (ARMORACIA RUSTICANA).
D'auria M, et al.
SPME-GC-MS ANALYSIS OF HORSERADISH (ARMORACIA RUSTICANA, 16(4) (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service