Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

16214

Supelco

Solvent Green 7 hydrate

analytical standard

Synonym(s):

8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt hydrate, D&C Green No. 8, Pyranine Green, HPTS, Pyranine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H7Na3O10S3 · xH2O
CAS Number:
Molecular Weight:
524.39 (anhydrous basis)
Colour Index Number:
59040
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

~12% water

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC1=C2C(C3=C(C=C4)C(S(=O)([O-])=O)=CC(S(=O)([O-])=O)=C3C=C2)=C4C(S(=O)([O-])=O)=C1.O.[Na+].[Na+].[Na+]

InChI

1S/C16H10O10S3.3Na.H2O/c17-11-5-12(27(18,19)20)8-3-4-10-14(29(24,25)26)6-13(28(21,22)23)9-2-1-7(11)15(8)16(9)10;;;;/h1-6,17H,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;;1H2/q;3*+1;/p-3

InChI key

DCEGPALHGHQUSK-UHFFFAOYSA-K

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

481.3 °F

Flash Point(C)

249.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Torre et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(31), 15378-15385 (2019-07-17)
Reconstructing the functions of living cells using nonnatural components is one of the great challenges of natural sciences. Compartmentalization, encapsulation, and surface decoration of globular assemblies, known as vesicles, represent key early steps in the reconstitution of synthetic cells. Here
Mauricio A García et al.
Pharmaceutical research, 37(7), 121-121 (2020-06-10)
MDCK cells are commonly used to assess drug permeability, but the existence of various strains merits a comparative functional study. Since metformin absorption is largely mediated by transporters and paracellular diffusion, we used it to functionally compare MDCK-wt and MDCK-II.
Aaron C Schmidt et al.
Soft matter, 16(11), 2725-2735 (2020-03-03)
Transmembrane pH gradient poly(isoprene)-block-poly(ethylene glycol) (PI-b-PEG) polymersomes were investigated for their potential use in the detoxification of ammonia, a metabolite that is excessively present in patients suffering from urea cycle disorders and advanced liver diseases, and which causes neurotoxic effects
Xie Dang et al.
Cell reports, 30(11), 3904-3916 (2020-03-19)
The flowers of angiosperm species typically contain specialized conical cells. Although substantial progress has been achieved regarding the mechanisms underlying flower development, little is known about how petal cells achieve final conical shape. Here, we use 8-hydroxypyrene-1,3,6-trisulfonic acid trisodium salt

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service