Skip to Content
MilliporeSigma
All Photos(1)

Documents

15859

Sigma-Aldrich

Bradykinin triacetate salt

PeptiSelect, ≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H73N15O11 · 3C2H4O2
CAS Number:
Molecular Weight:
1240.36
Beilstein:
5497442
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

PeptiSelect

Assay

≥96.0% (HPLC)

storage temp.

−20°C

SMILES string

N[C@@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)N2CCC[C@H]2C(=O)NCC(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CO)C(=O)N4CCC[C@H]4C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Other Notes

General review

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacology of bradykinin and related kinins.
D Regoli et al.
Pharmacological reviews, 32(1), 1-46 (1980-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service