Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15800

Sigma-Aldrich

Boron trifluoride methyl etherate

technical, ~60% BF3 basis

Synonym(s):

(Dimethyl ether)trifluoroboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · O(CH3)2
CAS Number:
Molecular Weight:
113.87
Beilstein:
3613739
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

technical

autoignition temp.

455 °F

expl. lim.

21.6 %

concentration

~60% (BF3)

refractive index

n20/D 1.308 (lit.)

bp

126-127 °C (lit.)

mp

−15 °C (lit.)

density

1.239 g/mL at 25 °C (lit.)

SMILES string

C[O+](C)[B-](F)(F)F

InChI

1S/C2H6BF3O/c1-7(2)3(4,5)6/h1-2H3

InChI key

FOWZWMGLMYVYDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Deconvoluting the chemical reaction kinetics of platinum-silver ligand exchange
  • Synthesis of neurogenic ganglioside LLG-3 from starfish via glycosylation reactions
  • Synthesis of peloruside via lactonization and relay ring-closing metathesis cyclization
  • FRET for monitoring Pd-catalyzed nucleophilic allylic substitution reactions
  • Single molecuel imaging of fluorophore-tagged palldium(II) complexes
  • β-elimination and rearrangement reactions for synthesis of exocyclic vinyl ethers

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service