Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

15630

Sigma-Aldrich

(R)-(+)-Bornylamine

purum, ≥97.0% (NT)

Synonym(s):

(+)-Bornanamine, endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N
CAS Number:
Molecular Weight:
153.26
Beilstein:
2801831
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (NT)

form

solid

optical activity

[α]20/D +46±2°, c = 0.5% in ethanol

mp

160-163 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)[C@@H](N)C2

InChI

1S/C10H19N/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8H,4-6,11H2,1-3H3/t7-,8+,10+/m1/s1

InChI key

MDFWXZBEVCOVIO-WEDXCCLWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresia A Carreon et al.
Scientific reports, 7(1), 452-452 (2017-03-30)
In the eye, intraocular pressure (IOP) is tightly regulated and its persistent increase leads to ocular hypertension and glaucoma. We have previously shown that trabecular meshwork (TM) cells might detect aqueous humor fluid shear stress via interaction of the extracellular
Peter Barbuti et al.
Cells, 9(9) (2020-09-16)
The generation of isogenic induced pluripotent stem cell (iPSC) lines using CRISPR-Cas9 technology is a technically challenging, time-consuming process with variable efficiency. Here we use fluorescence-activated cell sorting (FACS) to sort biallelic CRISPR-Cas9 edited single-cell iPSC clones into high-throughput 96-well
Daniel J Smit et al.
Cells, 9(9) (2020-09-24)
Circulating tumor cells (CTCs) are cells shed from the primary tumor into the bloodstream. While many studies on solid tumor cells exist, data on CTCs are scarce. The mortality of cancer is mostly associated with metastasis and recent research identified
Tânia D F Costa et al.
Nature communications, 10(1), 3589-3589 (2019-08-11)
Overcoming cellular growth restriction, including the evasion of cellular senescence, is a hallmark of cancer. We report that PAK4 is overexpressed in all human breast cancer subtypes and associated with poor patient outcome. In mice, MMTV-PAK4 overexpression promotes spontaneous mammary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service