Skip to Content
MilliporeSigma
All Photos(2)

Documents

14949

Supelco

5-Methyl-1H-benzotriazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
116658
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

210-212 °C/12 mmHg (lit.)

mp

80-82 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ccc2[nH]nnc2c1

InChI

1S/C7H7N3/c1-5-2-3-6-7(4-5)9-10-8-6/h2-4H,1H3,(H,8,9,10)

InChI key

LRUDIIUSNGCQKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Limited shelf life, expiry date on the label.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandros G Asimakopoulos et al.
Analytical chemistry, 85(1), 441-448 (2012-12-06)
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are used in a wide variety of industrial and consumer products and have been reported to occur in the environment. Owing to a lack of analytical methods, human exposure to BTR and BTH is
Devon A Cancilla et al.
Environmental toxicology and chemistry, 22(1), 134-140 (2002-12-31)
This paper presents the results of a number of field and laboratory studies to evaluate the environmental impact of aircraft deicing and anti-icing fluids (ADAFs) on aquatic systems. Both 5-methyl-1H-benzotriazole (5-MeBt) and 4-methyl-1H-benzotriazole (4-MeBt), known additives to ADAFs, were found
Attilio Naccarato et al.
Journal of chromatography. A, 1338, 164-173 (2014-03-19)
This work proposes a new approach for the simultaneous determination of benzothiazoles, benzotriazoles and benzosulfonamides in different environmental matrices and human urine, using solid-phase microextraction coupled with gas-chromatography-triple quadrupole mass spectrometry (SPME-GC-QqQMS). The analytes object of this investigation have been
Rozanna Avagyan et al.
Environmental science and pollution research international, 22(8), 5842-5849 (2014-10-25)
Textiles play an important role in our daily life, and textile production is one of the oldest industries. In the manufacturing chain from natural and/or synthetic fibers to the final clothing products, the use of many different chemicals is ubiquitous.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service