Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14470

Sigma-Aldrich

Zinc borate

technical, light, ≥45% ZnO basis, ≥36% B2O3 basis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

technical

form

solid

composition

B2O3, ≥36%
ZnO, ≥45%

reaction suitability

core: zinc
reagent type: catalyst

loss

≤19% loss on ignition, 800 °C, 1h

SMILES string

[Zn++].[Zn++].[Zn++].[O-]B([O-])[O-].[O-]B([O-])[O-]

InChI

1S/2BO3.3Zn/c2*2-1(3)4;;;/q2*-3;3*+2

InChI key

BIKXLKXABVUSMH-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. xrd data for zinc borate

    1 answer
    1. X-Ray diffraction analysis is not performed on this product. It may be helpful to view the peer-reviewed articles related to this product: https://www.sigmaaldrich.com/search/14470?focus=papers&page=1&perpage=30&sort=relevance&term=14470&type=citation_search

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service