Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14385

Sigma-Aldrich

Biochanin A

≥97.0% (TLC)

Synonym(s):

4′-Methylgenistein, 5,7-Dihydroxy-4′-methoxyisoflavone, Genistein 4′-methyl ether, Olmelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
278107
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥97.0% (TLC)

mp

210-213 °C (lit.)
212-218 °C

solubility

acetone: 10 mg/mL, clear, brown

SMILES string

COc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C16H12O5/c1-20-11-4-2-9(3-5-11)12-8-21-14-7-10(17)6-13(18)15(14)16(12)19/h2-8,17-18H,1H3

InChI key

WUADCCWRTIWANL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Biochanin A is an isoflavone phytoestrogen found in red clover (Trifolium pratense) that is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels.
Biochanin A is an isoflavone phytoestrogen found in red clover (Trifolium pratense) that is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels. Induction of sulfotransferases for xenobiotic detoxification has been proposed as a mechanism of its cancer preventive effects.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bárbara Socas-Rodríguez et al.
Journal of chromatography. A, 1496, 58-67 (2017-04-02)
In this work, a simple and fast methodology has been validated and applied for the analysis of a group of 22 estrogenic compounds including eight phytoestrogens (i.e. daidzein, enterodiol, glycitein, enterolactone, genistein, formononetin, prunetin, biochanin A), six mycotoxins (β-zearalanol, β-zearalenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service