Skip to Content
MilliporeSigma
All Photos(1)

Documents

13540

Sigma-Aldrich

Benzyl mercaptan

purum, ≥99.0% (GC)

Synonym(s):

α-Toluenethiol, α-Tolyl mercaptan, Benzenemethanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SH
CAS Number:
Molecular Weight:
124.20
Beilstein:
605864
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

>4 (vs air)

grade

purum

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.575 (lit.)
n20/D 1.576

bp

194-195 °C (lit.)

density

1.058 g/mL at 25 °C (lit.)

SMILES string

SCc1ccccc1

InChI

1S/C7H8S/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

UENWRTRMUIOCKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by B25401-ALDRICH | Benzyl mercaptan 99%

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minglu Gao et al.
PLoS pathogens, 16(7), e1008595-e1008595 (2020-07-07)
Sarocladium zeae is a fungal endophyte of maize and can be found co-inhabiting a single seed with Fusarium verticillioides, a major mycotoxigenic food safety threat. S. zeae produces pyrrocidines A and B that inhibit the growth of F. verticillioides and
Theresa K Tiefenbrunn et al.
Protein science : a publication of the Protein Society, 18(5), 970-979 (2009-04-23)
The type 1 repeat domain from thrombospondin has potent antiangiogenic activity and a structurally interesting fold, making it an attractive target for protein engineering. Chemical synthesis is an attractive approach for studying protein domains because it enables the use of
Hai-Chao Zhou et al.
PloS one, 9(10), e107606-e107606 (2014-10-15)
Tannins from the leaves of a medicinal mangrove plant, Ceriops tagal, were purified and fractionated on Sephadex LH-20 columns. 13C nuclear magnetic resonance (13C-NMR), reversed/normal high performance liquid chromatography electrospray ionization mass spectrometry (HPLC-ESI MS) and matrix-assisted laser desorption/ionization time-of-flight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service