Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13401

Sigma-Aldrich

Benzyldimethyltetradecylammonium chloride

anhydrous, ≥99.0% (AT)

Synonym(s):

Benzyltetradecyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
368.04
Beilstein:
4062599
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

description

cationic

Assay

≥99.0% (AT)

form

powder

quality

anhydrous

mp

56-62 °C

solubility

water: soluble 1 g/10 mL, clear, colorless

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1

InChI key

OCBHHZMJRVXXQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzyldimethyltetradecylammonium chloride (BTDACl), a quaternary ammonium compound, is a cationic surfactant. It is utilized as phase transfer catalyst in organic reactions. Kinetics, thermodynamics and reaction mechanism of biosorption of benzyldimethyltetradecylammonium chloride (tetradecyl benzyl dimethyl ammonium chloride) onto activated sludge has been studied.

Application

Benzyldimethyltetradecylammonium chloride (Dimethylbenzyltetradecylammonium chloride) may be used in the following studies:
  • Spectrophotometric determination of cobalt(II).
  • Preparation of mixed micelle with tetradecyltrimethylammonium (TTAB) and tetradecyltriphenylphosphonium bromides.
  • Preparation of modified analcime zeolite, used in the trace determination of cadmium.
  • As a phase transfer catalyst in the preparation of 2,3-epoxy-1-phenyl-3-aryl-1-propanones.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuru Hoshino et al.
Chemical & pharmaceutical bulletin, 59(6), 721-724 (2011-06-02)
Spectrophotometric determination of cobalt(II) was accomplished with vanillilfluorone (VF) in the presence of dimethylbenzyltetradecylammonium chloride (Zephiramine, Zep). In the determination of cobalt(II), Beer's law was obeyed in the range of 24-470 ng/ml, with an effective molar absorption coefficient (at 575
Rong Ren et al.
Bioresource technology, 102(4), 3799-3804 (2011-01-05)
The biosorption of tetradecyl benzyl dimethyl ammonium chloride (C(14)BDMA) onto activated sludge was examined in aqueous solution with respect to the contact time, temperature and particle size. Equilibrium reached in about 2h contact time. An decrease in the temperature increases
Patrick Chavant et al.
FEMS microbiology letters, 236(2), 241-248 (2004-07-15)
This study was designed to investigate the individual or combined effects of sanitizers on survival of planktonic or sessile Listeria monocytogenes cells at different phase of growth. The sanitizers tested included: (i) acetic acid (pH 5.0), (ii) NaOH (pH 12.0)
Synthesis of 2,3-epoxy-1-phenyl-3-aryl-1-propanone by combination of phase transfer catalyst and ultrasound irradiation.
Li JT, et al.
Organic Communications, 2, 1-6 (2009)
Y-Q Zhu et al.
The British journal of surgery, 100(6), 784-793 (2013-04-05)
Benign strictures at the cardia are troublesome for patients and often require repeated endoscopic treatments. Paclitaxel can reduce fibrosis. This study evaluated a biodegradable paclitaxel-eluting nanofibre-covered metal stent for the treatment of benign cardia stricture in vitro and in vivo.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service