Skip to Content
MilliporeSigma
All Photos(1)

Documents

13013

Sigma-Aldrich

Lithium chloride

puriss., anhydrous, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352300

Quality Level

grade

puriss.

Assay

≥98%

impurities

≤0.001% heavy metals (as Pb)

mp

605 °C (lit.)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤100 mg/kg
Fe: ≤10 mg/kg
K: ≤100 mg/kg
Na: ≤200 mg/kg

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amitava Adhikary et al.
Journal of the American Chemical Society, 131(24), 8614-8619 (2009-05-28)
In this work, it is shown that the incorporation of an 8-deuteroguanine (G*) moiety in DNA-oligomers allows for direct determination at 77 K of (i) the location of holes (i.e., the radical site) within dsDNA at specific base sites, even
Steven Zukerman et al.
Chemical senses, 34(8), 685-694 (2009-09-09)
Prior work has shown that sweet taste-deficient T1R3 knockout (KO) mice developed significant sucrose preferences when given long-term sugar versus water tests. The current study investigated the role of olfaction in this experience-conditioned sucrose preference. T1R3 KO and C57BL/6 wild-type
Kim Zarse et al.
European journal of nutrition, 50(5), 387-389 (2011-02-09)
Lithium is a nutritionally essential trace element predominantly contained in vegetables, plant-derived foods, and drinking water. Environmental lithium exposure and concurrent nutritional intake vary considerably in different regions. We here have analyzed the possibility that low-dose lithium exposure may affect
Anvita Kumar et al.
Journal of virology, 82(6), 2813-2820 (2008-01-18)
The Wnt/beta-catenin pathway is involved in cell functions governing development and disease. In modeling postentry restriction of human immunodeficiency virus (HIV) replication in astrocytes, we reported that part of this natural resistance to productive replication of HIV in astrocytes involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service